Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCGCACAGTCAAGTGCGTTAAC[C/T]GCAATCTGACCGAGGTGCCCACGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 190920 | ||||||||||||||||||||
Literature Links: |
TPBG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPBG - trophoblast glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166392.1 | 663 | Missense Mutation | CGC,TGC | R,C 80 | NP_001159864.1 | |
NM_006670.4 | 663 | Missense Mutation | CGC,TGC | R,C 80 | NP_006661.1 | |
XM_011536097.2 | 663 | Missense Mutation | CGC,TGC | R,C 80 | XP_011534399.1 |