Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAAAGAAATGGCTGCAAGCAAC[C/T]GACCTCACTAGAGAAGTGTACCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609134 | ||||||||||||||||||||
Literature Links: |
UBR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBR2 - ubiquitin protein ligase E3 component n-recognin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184801.1 | 394 | Silent Mutation | ACC,ACT | T,T 32 | NP_001171730.1 | |
NM_015255.2 | 394 | Silent Mutation | ACC,ACT | T,T 32 | NP_056070.1 | |
XM_005248965.4 | 394 | Silent Mutation | ACC,ACT | T,T 32 | XP_005249022.1 | |
XM_005248966.3 | 394 | Intron | XP_005249023.1 | |||
XM_011514438.2 | 394 | Silent Mutation | ACC,ACT | T,T 59 | XP_011512740.2 | |
XM_011514439.1 | 394 | Intron | XP_011512741.1 | |||
XM_011514440.1 | 394 | Intron | XP_011512742.1 | |||
XM_011514441.2 | 394 | Intron | XP_011512743.1 | |||
XM_017010594.1 | 394 | Silent Mutation | ACC,ACT | T,T 59 | XP_016866083.1 | |
XM_017010595.1 | 394 | Silent Mutation | ACC,ACT | T,T 59 | XP_016866084.1 | |
XM_017010596.1 | 394 | Silent Mutation | ACC,ACT | T,T 32 | XP_016866085.1 | |
XM_017010597.1 | 394 | Silent Mutation | ACC,ACT | T,T 59 | XP_016866086.1 |