Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCAGGCCGCGCAAGCAGCCTCC[A/G]GTGAGTCCCGGGACAGCGCTGGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611419 MIM: 600701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf1 - chromosome 6 open reading frame 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMGA1 - high mobility group AT-hook 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319077.1 | 591 | Silent Mutation | CCA,CCG | P,P 34 | NP_001306006.1 | |
NM_001319078.1 | 591 | Intron | NP_001306007.1 | |||
NM_001319079.1 | 591 | Silent Mutation | CCA,CCG | P,P 34 | NP_001306008.1 | |
NM_001319080.1 | 591 | Silent Mutation | CCA,CCG | P,P 34 | NP_001306009.1 | |
NM_001319081.1 | 591 | Intron | NP_001306010.1 | |||
NM_001319082.1 | 591 | Silent Mutation | CCA,CCG | P,P 34 | NP_001306011.1 | |
NM_002131.3 | 591 | Silent Mutation | CCA,CCG | P,P 34 | NP_002122.1 | |
NM_145899.2 | 591 | Silent Mutation | CCA,CCG | P,P 34 | NP_665906.1 | |
NM_145901.2 | 591 | Silent Mutation | CCA,CCG | P,P 34 | NP_665908.1 | |
NM_145902.2 | 591 | Silent Mutation | CCA,CCG | P,P 34 | NP_665909.1 | |
NM_145903.2 | 591 | Intron | NP_665910.1 | |||
NM_145905.2 | 591 | Intron | NP_665912.1 |
MIR6835 - microRNA 6835 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |