Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGAGGCCAGAGTGCCTAGAGCAC[A/G]GTGGAGGTAGTTGAGAAGATTGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608304 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAGE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAGE1 - cancer antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170692.1 | Intron | NP_001164163.1 | ||||
NM_001170693.1 | Intron | NP_001164164.1 | ||||
NM_205864.2 | Intron | NP_995586.1 |
RIOK1 - RIO kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031480.2 | Intron | NP_113668.2 | ||||
NM_153005.1 | Intron | NP_694550.1 | ||||
XM_005249446.3 | Intron | XP_005249503.1 | ||||
XM_011514933.2 | Intron | XP_011513235.2 |