Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCATCGATTAGCAGAAGAGTCC[A/G]GGACAAACGCTTGTGCGAGTAAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611264 | ||||||||||||||||||||
Literature Links: |
CENPW PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPW - centromere protein W | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012507.3 | 713 | Missense Mutation | AGG,GGG | R,G 59 | NP_001012525.1 | |
NM_001286524.1 | 713 | Missense Mutation | AGG,GGG | R,G 74 | NP_001273453.1 | |
NM_001286525.1 | 713 | Silent Mutation | CCA,CCG | P,P 45 | NP_001273454.1 | |
XM_017010845.1 | 713 | Silent Mutation | CCA,CCG | P,P 45 | XP_016866334.1 |