Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAGGAAGGTGAGCTGGACCACA[C/T]CCACTGGGGAGCCCTCTGAGTCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120290 | ||||||||||||||||||||
Literature Links: |
COL11A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL11A2 - collagen type XI alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163771.1 | 4417 | Intron | NP_001157243.1 | |||
NM_080679.2 | 4417 | Missense Mutation | GAT,GGT | D,G 1526 | NP_542410.2 | |
NM_080680.2 | 4417 | Missense Mutation | GAT,GGT | D,G 1633 | NP_542411.2 | |
NM_080681.2 | 4417 | Missense Mutation | GAT,GGT | D,G 1547 | NP_542412.2 | |
XM_011514299.2 | 4417 | Missense Mutation | GAT,GGT | D,G 1395 | XP_011512601.1 | |
XM_011514300.2 | 4417 | Missense Mutation | GAT,GGT | D,G 1335 | XP_011512602.1 | |
XM_011514302.2 | 4417 | Missense Mutation | GAT,GGT | D,G 1262 | XP_011512604.1 | |
XM_017010250.1 | 4417 | Missense Mutation | GAT,GGT | D,G 1633 | XP_016865739.1 | |
XM_017010251.1 | 4417 | Missense Mutation | GAT,GGT | D,G 1239 | XP_016865740.1 |