Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTGCTTTTGTTCATATATCTT[C/T]TGCACAATTTGAACTGTCACAAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604092 | ||||||||||||||||||||
Literature Links: |
TTK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTK - TTK protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166691.1 | 557 | Missense Mutation | TCT,TTT | S,F 149 | NP_001160163.1 | |
NM_003318.4 | 557 | Missense Mutation | TCT,TTT | S,F 149 | NP_003309.2 | |
XM_011536099.2 | 557 | Missense Mutation | TCT,TTT | S,F 149 | XP_011534401.1 | |
XM_011536100.2 | 557 | Missense Mutation | TCT,TTT | S,F 149 | XP_011534402.1 | |
XM_017011242.1 | 557 | Missense Mutation | TCT,TTT | S,F 94 | XP_016866731.1 |