Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGTATGGCTGAGTGAGGTGGG[C/T]GCTGGGTGCAGGGCCAGCTGGTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FRMD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRMD1 - FERM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122841.2 | 1412 | Silent Mutation | GCA,GCG | A,A 431 | NP_001116313.1 | |
NM_024919.4 | 1412 | Silent Mutation | GCA,GCG | A,A 499 | NP_079195.3 | |
XM_011536137.1 | 1412 | Silent Mutation | GCA,GCG | A,A 564 | XP_011534439.1 | |
XM_011536138.1 | 1412 | Silent Mutation | GCA,GCG | A,A 502 | XP_011534440.1 | |
XM_011536140.1 | 1412 | Silent Mutation | GCA,GCG | A,A 475 | XP_011534442.1 | |
XM_011536141.1 | 1412 | Silent Mutation | GCA,GCG | A,A 456 | XP_011534443.1 | |
XM_011536142.2 | 1412 | Silent Mutation | GCA,GCG | A,A 456 | XP_011534444.1 | |
XM_011536143.1 | 1412 | Silent Mutation | GCA,GCG | A,A 456 | XP_011534445.1 | |
XM_011536145.1 | 1412 | Silent Mutation | GCA,GCG | A,A 454 | XP_011534447.1 | |
XM_011536146.2 | 1412 | Silent Mutation | GCA,GCG | A,A 293 | XP_011534448.1 | |
XM_017011317.1 | 1412 | Silent Mutation | GCA,GCG | A,A 541 | XP_016866806.1 | |
XM_017011318.1 | 1412 | Silent Mutation | GCA,GCG | A,A 293 | XP_016866807.1 |