Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTCGTAGTCGGCATGCAACCAT[A/G]AGAAGCAGCAGAGCAGTGATAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP11 - LDL receptor related protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032832.5 | 1725 | Silent Mutation | CTC,CTT | L,L 467 | NP_116221.3 | |
XM_011536194.2 | 1725 | Silent Mutation | CTC,CTT | L,L 435 | XP_011534496.1 | |
XM_017011380.1 | 1725 | Intron | XP_016866869.1 |
PCMT1 - protein-L-isoaspartate (D-aspartate) O-methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |