Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTTTGACATAGGGGCTAGTCTC[C/T]GGGCTCAGCTCATCCTCATTGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL11A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL11A2 - collagen type XI alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163771.1 | 4547 | Intron | NP_001157243.1 | |||
NM_080679.2 | 4547 | Silent Mutation | CCA,CCG | P,P 1569 | NP_542410.2 | |
NM_080680.2 | 4547 | Silent Mutation | CCA,CCG | P,P 1676 | NP_542411.2 | |
NM_080681.2 | 4547 | Silent Mutation | CCA,CCG | P,P 1590 | NP_542412.2 | |
XM_011514299.2 | 4547 | Silent Mutation | CCA,CCG | P,P 1438 | XP_011512601.1 | |
XM_011514300.2 | 4547 | Silent Mutation | CCA,CCG | P,P 1378 | XP_011512602.1 | |
XM_011514302.2 | 4547 | Silent Mutation | CCA,CCG | P,P 1305 | XP_011512604.1 | |
XM_017010250.1 | 4547 | Silent Mutation | CCA,CCG | P,P 1676 | XP_016865739.1 | |
XM_017010251.1 | 4547 | Silent Mutation | CCA,CCG | P,P 1282 | XP_016865740.1 |