Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGTGAATTTTTTGAGAAACACG[A/G]AGAGGTAAGAATTGTTTTAACTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616332 MIM: 614917 | ||||||||||||||||||||
Literature Links: |
ARMT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMT1 - acidic residue methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286562.1 | 431 | Intron | NP_001273491.1 | |||
NM_024573.2 | 431 | Missense Mutation | GAA,GGA | E,G 48 | NP_078849.1 |
RMND1 - required for meiotic nuclear division 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |