Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCATATTATGGGTGATAAGGTC[C/T]CAGATTAACGTAAGAAAACGAGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612588 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCLAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCLAF1 - BCL2 associated transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077440.1 | 6026 | UTR 3 | NP_001070908.1 | |||
NM_001077441.1 | 6026 | UTR 3 | NP_001070909.1 | |||
NM_001301038.1 | 6026 | UTR 3 | NP_001287967.1 | |||
NM_014739.2 | 6026 | UTR 3 | NP_055554.1 | |||
XM_005267237.3 | 6026 | Intron | XP_005267294.1 | |||
XM_017011529.1 | 6026 | UTR 3 | XP_016867018.1 | |||
XM_017011530.1 | 6026 | UTR 3 | XP_016867019.1 | |||
XM_017011531.1 | 6026 | UTR 3 | XP_016867020.1 | |||
XM_017011532.1 | 6026 | UTR 3 | XP_016867021.1 |
MTFR2 - mitochondrial fission regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |