Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCTAGCAACAACTGACAGGCA[C/G]CCAGATTGGCATGAAGGACAGTTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600984 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATF6B - activating transcription factor 6 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136153.1 | 1046 | Intron | NP_001129625.1 | |||
NM_004381.4 | 1046 | Intron | NP_004372.3 |
FKBPL - FK506 binding protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022110.3 | 1046 | Missense Mutation | GCT,GGT | A,G 259 | NP_071393.2 |