Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGCTAACTTCGGTTCATCAAAG[C/T]AGTGACTGAAAAAAAGGTAGTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606117 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYRM4-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYRM4-AS1 - LYRM4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPP40 - ribonuclease P/MRP subunit p40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286132.1 | 834 | Missense Mutation | TAC,TGC | Y,C 277 | NP_001273061.1 | |
NM_001286133.1 | 834 | Missense Mutation | TAC,TGC | Y,C 258 | NP_001273062.1 | |
NM_006638.3 | 834 | Missense Mutation | TAC,TGC | Y,C 300 | NP_006629.2 | |
XM_011514253.2 | 834 | Intron | XP_011512555.1 |