Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCAAGCAGAAGAAAGGCAGACA[A/G]GAAAAGGCCTACGCTGTCCTCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602306 MIM: 601962 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGL2 - ral guanine nucleotide dissociation stimulator like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAPBP - TAP binding protein (tapasin) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003190.4 | 1465 | Silent Mutation | CTG,TTG | L,L 418 | NP_003181.3 | |
NM_172208.2 | 1465 | Silent Mutation | CTG,TTG | L,L 418 | NP_757345.2 | |
NM_172209.2 | 1465 | Silent Mutation | CTG,TTG | L,L 331 | NP_757346.2 | |
XM_011514828.2 | 1465 | Intron | XP_011513130.1 | |||
XM_017011227.1 | 1465 | Silent Mutation | CTG,TTG | L,L 418 | XP_016866716.1 |