Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGGGATGGACCTGGCTTTCTCA[A/G]CCAGGAATTTGTTATTGTGGCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612121 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNPLA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNPLA1 - patatin like phospholipase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145716.2 | Intron | NP_001139188.1 | ||||
NM_001145717.1 | Intron | NP_001139189.2 | ||||
NM_173676.2 | Intron | NP_775947.2 | ||||
XM_011514519.2 | Intron | XP_011512821.1 | ||||
XM_011514520.2 | Intron | XP_011512822.1 | ||||
XM_017010776.1 | Intron | XP_016866265.1 | ||||
XM_017010777.1 | Intron | XP_016866266.1 | ||||
XM_017010778.1 | Intron | XP_016866267.1 | ||||
XM_017010779.1 | Intron | XP_016866268.1 |