Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAACCATATCTTTTGCTCCTTG[G/T]TTCTTCTGATCTGTAGTTTACTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZKSCAN8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZKSCAN8 - zinc finger with KRAB and SCAN domains 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278119.1 | Intron | NP_001265048.1 | ||||
NM_001278121.1 | Intron | NP_001265050.1 | ||||
NM_001278122.1 | Intron | NP_001265051.1 | ||||
NM_006298.3 | Intron | NP_006289.2 | ||||
XM_011514870.2 | Intron | XP_011513172.1 | ||||
XM_011514873.2 | Intron | XP_011513175.1 | ||||
XM_011514874.2 | Intron | XP_011513176.1 | ||||
XM_017011266.1 | Intron | XP_016866755.1 |
ZSCAN16-AS1 - ZSCAN16 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |