Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTTGATAATGATCCCCCGGGG[A/G]CCAGGAGCTGCATAGGCCACAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120105 MIM: 609427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLPS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLPS - colipase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252597.1 | 116 | UTR 5 | NP_001239526.1 | |||
NM_001252598.1 | 116 | Silent Mutation | GGC,GGT | G,G 20 | NP_001239527.1 | |
NM_001832.3 | 116 | Silent Mutation | GGC,GGT | G,G 20 | NP_001823.1 |
CLPSL1 - colipase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LHFPL5 - lipoma HMGIC fusion partner-like 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |