Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613927 MIM: 138470 MIM: 154040 | ||||||||||||||||||||
Literature Links: |
C2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2 - complement component 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C2-AS1 - C2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CFB - complement factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001710.5 | 1033 | Missense Mutation | GAC,GGC | D,G 252 | NP_001701.2 |
MIR1236 - microRNA 1236 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NELFE - negative elongation factor complex member E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |