Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGTAGAAGCAGCAGTCATAAG[A/G]TCTAACATTTATTGAGCCTGTACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602209 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RREB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RREB1 - ras responsive element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003698.3 | Intron | NP_001003698.1 | ||||
NM_001003699.3 | Intron | NP_001003699.1 | ||||
NM_001003700.1 | Intron | NP_001003700.1 | ||||
NM_001168344.1 | Intron | NP_001161816.1 | ||||
XM_006715157.3 | Intron | XP_006715220.1 | ||||
XM_011514794.2 | Intron | XP_011513096.1 | ||||
XM_011514795.2 | Intron | XP_011513097.1 |