Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCCACCTACTGTGGTCTGACAG[C/T]GCAATATCCTGGTAAGAGAATTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPX5 - glutathione peroxidase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001509.2 | 232 | Missense Mutation | GCG,GTG | A,V 77 | NP_001500.1 | |
NM_003996.3 | 232 | Missense Mutation | GCG,GTG | A,V 77 | NP_003987.2 |