Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAATTTCAACCGCTCTGTGAGC[C/T]CCTTCATAAACCTTTTGCTCCTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606648 | ||||||||||||||||||||
Literature Links: |
IL22RA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL22RA2 - interleukin 22 receptor subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052962.2 | 868 | Silent Mutation | GGA,GGG | G,G 221 | NP_443194.1 | |
NM_181309.1 | 868 | Silent Mutation | GGA,GGG | G,G 189 | NP_851826.1 | |
NM_181310.1 | 868 | UTR 3 | NP_851827.1 |