Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCAGGCCCTTTGTTAAATGCTC[A/G]CTGACCAGCCTGCATGGTGCAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120165 MIM: 120210 | ||||||||||||||||||||
Literature Links: |
COL19A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL19A1 - collagen type XIX alpha 1 chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COL9A1 - collagen type IX alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001851.4 | 2283 | Silent Mutation | NP_001842.3 | |||
NM_078485.3 | 2283 | Silent Mutation | NP_511040.2 | |||
XM_011535429.2 | 2283 | Nonsense Mutation | XP_011533731.1 | |||
XM_011535430.2 | 2283 | Silent Mutation | XP_011533732.1 | |||
XM_017010246.1 | 2283 | Nonsense Mutation | XP_016865735.1 | |||
XM_017010247.1 | 2283 | Nonsense Mutation | XP_016865736.1 |