Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTAAGATGCCATGGCTAGTCC[A/G]GGGAAAGATAACTATAGAATGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604545 | ||||||||||||||||||||
Literature Links: |
KPNA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KPNA5 - karyopherin subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002269.2 | 133 | Silent Mutation | CCA,CCG | P,P 7 | NP_002260.2 | |
XM_011535805.1 | 133 | Silent Mutation | CCA,CCG | P,P 27 | XP_011534107.1 | |
XM_017010838.1 | 133 | Silent Mutation | CCA,CCG | P,P 27 | XP_016866327.1 | |
XM_017010839.1 | 133 | Silent Mutation | CCA,CCG | P,P 7 | XP_016866328.1 | |
XM_017010840.1 | 133 | Silent Mutation | CCA,CCG | P,P 4 | XP_016866329.1 | |
XM_017010841.1 | 133 | Intron | XP_016866330.1 | |||
XM_017010842.1 | 133 | UTR 5 | XP_016866331.1 | |||
XM_017010843.1 | 133 | UTR 5 | XP_016866332.1 |