Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGTTGGTCCCTGGAATAAGATT[A/C]TCATCATCTCCTTCAATCAAGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601690 MIM: 611200 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G7 - phospholipase A2 group VII | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168357.1 | 1420 | Missense Mutation | GAG,GAT | E,D 414 | NP_001161829.1 | |
NM_005084.3 | 1420 | Missense Mutation | GAG,GAT | E,D 414 | NP_005075.3 | |
XM_005249408.4 | 1420 | Missense Mutation | GAG,GAT | E,D 414 | XP_005249465.1 |
TDRD6 - tudor domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010870.2 | 1420 | Intron | NP_001010870.1 | |||
NM_001168359.1 | 1420 | Intron | NP_001161831.1 |