Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGATCTCTCAGCATACTTCACT[C/T]TGAGTTAATCAAAAATTTTCAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600211 MIM: 602947 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUNX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RUNX2 - runt related transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015051.3 | Intron | NP_001015051.3 | ||||
NM_001024630.3 | Intron | NP_001019801.3 | ||||
NM_001278478.1 | Intron | NP_001265407.1 | ||||
XM_006715232.1 | Intron | XP_006715295.1 | ||||
XM_011514960.2 | Intron | XP_011513262.1 | ||||
XM_011514961.2 | Intron | XP_011513263.1 | ||||
XM_011514962.2 | Intron | XP_011513264.1 | ||||
XM_011514963.2 | Intron | XP_011513265.1 | ||||
XM_011514964.2 | Intron | XP_011513266.1 | ||||
XM_011514965.2 | Intron | XP_011513267.1 | ||||
XM_011514966.2 | Intron | XP_011513268.1 | ||||
XM_017011391.1 | Intron | XP_016866880.1 | ||||
XM_017011392.1 | Intron | XP_016866881.1 | ||||
XM_017011393.1 | Intron | XP_016866882.1 | ||||
XM_017011394.1 | Intron | XP_016866883.1 | ||||
XM_017011395.1 | Intron | XP_016866884.1 | ||||
XM_017011396.1 | Intron | XP_016866885.1 |
SUPT3H - SPT3 homolog, SAGA and STAGA complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261823.1 | Intron | NP_001248752.1 | ||||
NM_003599.3 | Intron | NP_003590.1 | ||||
NM_181356.2 | Intron | NP_852001.1 | ||||
XM_011514949.2 | Intron | XP_011513251.1 | ||||
XM_011514950.1 | Intron | XP_011513252.1 | ||||
XM_011514951.2 | Intron | XP_011513253.1 | ||||
XM_011514952.2 | Intron | XP_011513254.1 | ||||
XM_011514953.2 | Intron | XP_011513255.1 | ||||
XM_011514954.2 | Intron | XP_011513256.1 | ||||
XM_017011369.1 | Intron | XP_016866858.1 | ||||
XM_017011370.1 | Intron | XP_016866859.1 | ||||
XM_017011371.1 | Intron | XP_016866860.1 | ||||
XM_017011372.1 | Intron | XP_016866861.1 | ||||
XM_017011373.1 | Intron | XP_016866862.1 | ||||
XM_017011374.1 | Intron | XP_016866863.1 |