Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAAAGAGAGTGTGATGACAGAA[C/T]ACCTTAGACTTCAACACGCATCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608313 MIM: 605042 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARG1 - arginase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000045.3 | 4463 | Intron | NP_000036.2 | |||
NM_001244438.1 | 4463 | Intron | NP_001231367.1 | |||
XM_011535801.2 | 4463 | Intron | XP_011534103.1 |
MED23 - mediator complex subunit 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270521.1 | 4463 | UTR 3 | NP_001257450.1 | |||
NM_001270522.1 | 4463 | Intron | NP_001257451.1 | |||
NM_004830.3 | 4463 | Intron | NP_004821.2 | |||
NM_015979.3 | 4463 | UTR 3 | NP_057063.2 | |||
XM_005267223.2 | 4463 | Intron | XP_005267280.1 | |||
XM_006715612.2 | 4463 | Intron | XP_006715675.1 | |||
XM_011536257.2 | 4463 | Intron | XP_011534559.1 | |||
XM_017011501.1 | 4463 | Intron | XP_016866990.1 |