Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTCCTGACTACAAGTCCTTTTG[C/T]CGTGAAGAGAAAGAATGGCCTCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606749 | ||||||||||||||||||||
Literature Links: |
TINAG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TINAG - tubulointerstitial nephritis antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014464.3 | 452 | Silent Mutation | TGC,TGT | C,C 102 | NP_055279.3 | |
XM_006715062.2 | 452 | Silent Mutation | TGC,TGT | C,C 84 | XP_006715125.1 | |
XM_011514497.2 | 452 | Silent Mutation | TGC,TGT | C,C 102 | XP_011512799.1 | |
XM_017010745.1 | 452 | Silent Mutation | TGC,TGT | C,C 102 | XP_016866234.1 | |
XM_017010746.1 | 452 | Intron | XP_016866235.1 | |||
XM_017010747.1 | 452 | Intron | XP_016866236.1 | |||
XM_017010748.1 | 452 | Intron | XP_016866237.1 |