Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCAAGGAAGATTTTCCTCCCCA[A/G]GTTATTCTCTGATGTGGCAGCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCML4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCML4 - sex comb on midleg-like 4 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286408.1 | 4286 | Intron | NP_001273337.1 | |||
NM_001286409.1 | 4286 | Intron | NP_001273338.1 | |||
NM_198081.4 | 4286 | Intron | NP_932347.2 | |||
XM_011535703.2 | 4286 | UTR 3 | XP_011534005.1 | |||
XM_011535704.2 | 4286 | UTR 3 | XP_011534006.1 | |||
XM_011535706.1 | 4286 | UTR 3 | XP_011534008.1 | |||
XM_011535707.2 | 4286 | Intron | XP_011534009.1 | |||
XM_011535709.2 | 4286 | UTR 3 | XP_011534011.1 | |||
XM_011535710.2 | 4286 | UTR 3 | XP_011534012.1 | |||
XM_017010677.1 | 4286 | UTR 3 | XP_016866166.1 | |||
XM_017010678.1 | 4286 | UTR 3 | XP_016866167.1 | |||
XM_017010679.1 | 4286 | UTR 3 | XP_016866168.1 | |||
XM_017010680.1 | 4286 | UTR 3 | XP_016866169.1 | |||
XM_017010681.1 | 4286 | UTR 3 | XP_016866170.1 | |||
XM_017010682.1 | 4286 | UTR 3 | XP_016866171.1 | |||
XM_017010683.1 | 4286 | UTR 3 | XP_016866172.1 | |||
XM_017010684.1 | 4286 | Intron | XP_016866173.1 | |||
XM_017010685.1 | 4286 | UTR 3 | XP_016866174.1 | |||
XM_017010686.1 | 4286 | UTR 3 | XP_016866175.1 | |||
XM_017010687.1 | 4286 | UTR 3 | XP_016866176.1 | |||
XM_017010688.1 | 4286 | Intron | XP_016866177.1 |