Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCAGCCAGTCTCTGTACCGCAC[C/G]TCCGGAGCTTTCGTCTACGACTGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601090 MIM: 615976 | ||||||||||||||||||||
Literature Links: |
FOXC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXC1 - forkhead box C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001453.2 | 1626 | Silent Mutation | ACC,ACG | T,T 542 | NP_001444.2 |
FOXCUT - FOXC1 upstream transcript (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |