Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTCAGCTTGTCGTAGTTCATGG[C/T]GGGACGGTTCTTGCGGATGCCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606512 MIM: 608144 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PACSIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PACSIN1 - protein kinase C and casein kinase substrate in neurons 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPDEF - SAM pointed domain containing ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252294.1 | 2021 | Missense Mutation | ACC,GCC | T,A 283 | NP_001239223.1 | |
NM_012391.2 | 2021 | Missense Mutation | ACC,GCC | T,A 299 | NP_036523.1 | |
XM_005248988.4 | 2021 | Missense Mutation | ACC,GCC | T,A 371 | XP_005249045.2 | |
XM_011514457.2 | 2021 | Intron | XP_011512759.1 |