Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGGTGGCTGAGCTGAGCCAGC[A/G]CTGTGGCTGGAGCCCTGCGGAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC22A1 - solute carrier family 22 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003057.2 | 329 | Missense Mutation | CAC,CGC | H,R 61 | NP_003048.1 | |
NM_153187.1 | 329 | Missense Mutation | CAC,CGC | H,R 61 | NP_694857.1 | |
XM_005267102.4 | 329 | Missense Mutation | CAC,CGC | H,R 61 | XP_005267159.1 | |
XM_005267103.1 | 329 | Missense Mutation | CAC,CGC | H,R 61 | XP_005267160.1 | |
XM_005267104.4 | 329 | Intron | XP_005267161.1 | |||
XM_005267105.4 | 329 | Intron | XP_005267162.1 | |||
XM_006715552.1 | 329 | Missense Mutation | CAC,CGC | H,R 61 | XP_006715615.1 | |
XM_011536074.2 | 329 | Intron | XP_011534376.1 |