Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGCCTCATTTGCCTCTCGCCT[A/C]TTTTCGACCACCATTTTGGGGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601760 MIM: 612802 | ||||||||||||||||||||
Literature Links: |
GTF2H4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTF2H4 - general transcription factor IIH subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001517.4 | 187 | Intron | NP_001508.1 |
VARS2 - valyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167733.2 | 187 | Intron | NP_001161205.1 | |||
NM_001167734.1 | 187 | Missense Mutation | TAT,TCT | Y,S 38 | NP_001161206.1 | |
NM_020442.5 | 187 | Missense Mutation | TAT,TCT | Y,S 8 | NP_065175.4 |