Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGACAACCTAAAGCTCAGGAT[A/G]AAGGGCGTTGAAGGTCCTTTTTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607977 MIM: 611438 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HECA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HECA - hdc homolog, cell cycle regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016217.2 | Intron | NP_057301.1 |
TXLNB - taxilin beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153235.3 | Intron | NP_694967.3 | ||||
XM_005266836.1 | Intron | XP_005266893.1 | ||||
XM_011535505.2 | Intron | XP_011533807.1 | ||||
XM_011535506.2 | Intron | XP_011533808.1 | ||||
XM_017010322.1 | Intron | XP_016865811.1 |