Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGATCATTTCATTCAAATCTGCC[A/G]TACCTTCTGTATACATATCAATTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605449 | ||||||||||||||||||||
Literature Links: |
GSTA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTA3 - glutathione S-transferase alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000847.4 | 201 | Missense Mutation | ACG,ATG | T,M 99 | NP_000838.3 | |
XM_006715072.3 | 201 | Missense Mutation | ACG,ATG | T,M 49 | XP_006715135.1 | |
XM_011514533.2 | 201 | Missense Mutation | ACG,ATG | T,M 6 | XP_011512835.1 | |
XM_017010794.1 | 201 | Missense Mutation | ACG,ATG | T,M 49 | XP_016866283.1 |