Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTGCCACAGAAGGGCAGGCAGA[A/T]TGTCAACACTGTATGCACCACTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR5V1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR5V1 - olfactory receptor family 5 subfamily V member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030876.5 | 747 | Missense Mutation | ATC,TTC | I,F 164 | NP_110503.3 | |
XM_011514929.2 | 747 | Missense Mutation | ATC,TTC | I,F 164 | XP_011513231.1 | |
XM_011514930.2 | 747 | Missense Mutation | ATC,TTC | I,F 164 | XP_011513232.1 |