Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATCTGCCTCCTGGACTTCGAG[C/T]GGCACCTTGACAGGATAGCCCGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192150 MIM: 609693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VARS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VARS - valyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006295.2 | 4115 | Silent Mutation | CCA,CCG | P,P 1232 | NP_006286.1 | |
XM_005249362.2 | 4115 | Silent Mutation | CCA,CCG | P,P 1233 | XP_005249419.1 | |
XM_017011246.1 | 4115 | Silent Mutation | CCA,CCG | P,P 650 | XP_016866735.1 | |
XM_017011247.1 | 4115 | Silent Mutation | CCA,CCG | P,P 650 | XP_016866736.1 |
VWA7 - von Willebrand factor A domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |