Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGTTGGGTAGCCGAATATCATA[A/G]TGATATCTTTTTCCTATATATACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605824 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BVES-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BVES-AS1 - BVES antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POPDC3 - popeye domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022361.4 | 2076 | Silent Mutation | CAC,CAT | H,H 257 | NP_071756.2 | |
XM_011536067.2 | 2076 | Silent Mutation | CAC,CAT | H,H 257 | XP_011534369.1 | |
XM_017011194.1 | 2076 | Silent Mutation | CAC,CAT | H,H 257 | XP_016866683.1 | |
XM_017011195.1 | 2076 | Silent Mutation | CAC,CAT | H,H 257 | XP_016866684.1 |