Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCGGCGTATAACAGAGATAGAA[A/G]AGGAAGATAAGTAATTGCAATGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL12A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL12A1 - collagen type XII alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004370.5 | 9866 | UTR 3 | NP_004361.3 | |||
NM_080645.2 | 9866 | UTR 3 | NP_542376.2 | |||
XM_011535434.1 | 9866 | UTR 3 | XP_011533736.1 | |||
XM_011535435.1 | 9866 | UTR 3 | XP_011533737.1 | |||
XM_011535436.2 | 9866 | UTR 3 | XP_011533738.1 | |||
XM_017010252.1 | 9866 | UTR 3 | XP_016865741.1 |