Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAGAGGCGGAGTGCAAATGATA[C/T]GAACAACGTCACTAGTTTTCTCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM38 - tripartite motif containing 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006355.4 | 31 | UTR 5 | NP_006346.1 | |||
XM_005248799.4 | 31 | Intron | XP_005248856.1 | |||
XM_005248800.4 | 31 | Intron | XP_005248857.1 | |||
XM_017010175.1 | 31 | Intron | XP_016865664.1 |