Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTAATGAGATTGCACTGGCTGC[A/T]TTCACAACGTCTGCAGCATTTATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR14J1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR14J1 - olfactory receptor family 14 subfamily J member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030946.1 | 597 | Silent Mutation | GCA,GCT | A,A 199 | NP_112208.1 |