Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTTCCATTGTTATGCAGATGC[C/T]GTTTTAGTCATCCTGGAAAGAAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614738 | ||||||||||||||||||||
Literature Links: |
MPC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPC1 - mitochondrial pyruvate carrier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270879.1 | 1238 | Silent Mutation | ACA,ACG | T,T 63 | NP_001257808.1 | |
NM_016098.3 | 1238 | Silent Mutation | ACA,ACG | T,T 106 | NP_057182.1 | |
XM_011535894.1 | 1238 | Silent Mutation | ACA,ACG | T,T 63 | XP_011534196.1 | |
XM_011535895.1 | 1238 | Silent Mutation | ACA,ACG | T,T 63 | XP_011534197.1 |