Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCACTGAGCCAGCAAGATGGCAT[C/T]GAGCAGCACATGGATTTTGACAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607489 MIM: 600589 | ||||||||||||||||||||
Literature Links: |
CUL9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUL9 - cullin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015089.3 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | NP_055904.1 | |
XM_011514422.1 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | XP_011512724.1 | |
XM_011514423.2 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | XP_011512725.1 | |
XM_011514424.1 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | XP_011512726.1 | |
XM_011514425.1 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | XP_011512727.1 | |
XM_011514426.1 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | XP_011512728.1 | |
XM_011514427.1 | 1437 | Intron | XP_011512729.1 | |||
XM_011514428.1 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | XP_011512730.1 | |
XM_011514429.1 | 1437 | Intron | XP_011512731.1 | |||
XM_011514430.1 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | XP_011512732.1 | |
XM_017010589.1 | 1437 | Silent Mutation | ATC,ATT | I,I 214 | XP_016866078.1 | |
XM_017010590.1 | 1437 | Intron | XP_016866079.1 |
SRF - serum response factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |