Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTAATAGAGTTCAGAGTCTGAGG[A/C]CGCCTTTGCGTATGAGCTTGTGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611732 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBOAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBOAT1 - membrane bound O-acyltransferase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080480.2 | 1413 | Silent Mutation | NP_001073949.1 | |||
XM_006714999.2 | 1413 | Silent Mutation | XP_006715062.1 | |||
XM_006715000.3 | 1413 | Intron | XP_006715063.1 | |||
XM_011514313.2 | 1413 | Intron | XP_011512615.1 |