Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACCGGTGAGCACTCTGGGAATC[A/G]TGAGACACAGTAGAACTGAGCGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602448 | ||||||||||||||||||||
Literature Links: |
MAP3K5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K5 - mitogen-activated protein kinase kinase kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005923.3 | 3628 | Silent Mutation | CAC,CAT | H,H 1232 | NP_005914.1 | |
XM_011535839.2 | 3628 | Intron | XP_011534141.2 | |||
XM_017010870.1 | 3628 | Silent Mutation | CAC,CAT | H,H 1342 | XP_016866359.1 | |
XM_017010871.1 | 3628 | Silent Mutation | CAC,CAT | H,H 1341 | XP_016866360.1 | |
XM_017010872.1 | 3628 | Silent Mutation | CAC,CAT | H,H 1342 | XP_016866361.1 | |
XM_017010873.1 | 3628 | Silent Mutation | CAC,CAT | H,H 1341 | XP_016866362.1 | |
XM_017010874.1 | 3628 | Silent Mutation | CAC,CAT | H,H 1306 | XP_016866363.1 | |
XM_017010875.1 | 3628 | Silent Mutation | CAC,CAT | H,H 1305 | XP_016866364.1 | |
XM_017010876.1 | 3628 | Silent Mutation | CAC,CAT | H,H 1039 | XP_016866365.1 | |
XM_017010877.1 | 3628 | Silent Mutation | CAC,CAT | H,H 1039 | XP_016866366.1 |