Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142620 MIM: 606038 MIM: 610437 MIM: 611404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD16A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD16A - abhydrolase domain containing 16A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G6D - lymphocyte antigen 6 complex, locus G6D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G6E - lymphocyte antigen 6 complex, locus G6E (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G6F - lymphocyte antigen 6 complex, locus G6F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003693.1 | 495 | Silent Mutation | CCC,CCG | P,P 165 | NP_001003693.1 |
MIR4646 - microRNA 4646 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |