Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTCACAGATGAGGAGAAACGT[C/T]TGATGGTGGAGCTGCACAACCTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PI16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PI16 - peptidase inhibitor 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199159.1 | 326 | Silent Mutation | CTG,TTG | L,L 35 | NP_001186088.1 | |
NM_153370.2 | 326 | Silent Mutation | CTG,TTG | L,L 35 | NP_699201.2 | |
XM_005248917.2 | 326 | Silent Mutation | CTG,TTG | L,L 35 | XP_005248974.1 | |
XM_011514375.2 | 326 | Silent Mutation | CTG,TTG | L,L 35 | XP_011512677.1 | |
XM_017010430.1 | 326 | Silent Mutation | CTG,TTG | L,L 35 | XP_016865919.1 |