Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCAGCTTCCTTGACGCTGGCTGT[A/G]ACGAGGACGCTGTCAGGGGACTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602958 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGK1 - serum/glucocorticoid regulated kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143676.1 | 1355 | Silent Mutation | GTC,GTT | V,V 502 | NP_001137148.1 | |
NM_001143677.1 | 1355 | Silent Mutation | GTC,GTT | V,V 435 | NP_001137149.1 | |
NM_001143678.1 | 1355 | Silent Mutation | GTC,GTT | V,V 421 | NP_001137150.1 | |
NM_001291995.1 | 1355 | Silent Mutation | GTC,GTT | V,V 363 | NP_001278924.1 | |
NM_005627.3 | 1355 | Silent Mutation | GTC,GTT | V,V 407 | NP_005618.2 | |
XM_011536071.1 | 1355 | Silent Mutation | GTC,GTT | V,V 502 | XP_011534373.1 |