Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTGTTGGTCTCAGTCTGTTGTT[A/G]GCCTGGATGGAGACAAACTTGTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602965 MIM: 610728 | ||||||||||||||||||||
Literature Links: |
FABP7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FABP7 - fatty acid binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319039.1 | 317 | Missense Mutation | AGC,GGC | S,G 86 | NP_001305968.1 | |
NM_001319041.1 | 317 | UTR 3 | NP_001305970.1 | |||
NM_001319042.1 | 317 | Missense Mutation | AGC,GGC | S,G 82 | NP_001305971.1 | |
NM_001446.4 | 317 | Missense Mutation | AGC,GGC | S,G 86 | NP_001437.1 |
SMPDL3A - sphingomyelin phosphodiesterase acid like 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |