Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCGCTGTCTACCCCGACTCAGG[C/T]ACTCAGGTATGTCTCAAGTCCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRM1 - glutamate metabotropic receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278064.1 | 153 | Intron | NP_001264993.1 | |||
NM_001278065.1 | 153 | UTR 5 | NP_001264994.1 | |||
NM_001278066.1 | 153 | Intron | NP_001264995.1 | |||
NM_001278067.1 | 153 | Intron | NP_001264996.1 | |||
XM_011535782.1 | 153 | UTR 5 | XP_011534084.1 | |||
XM_017010783.1 | 153 | UTR 5 | XP_016866272.1 | |||
XM_017010784.1 | 153 | UTR 5 | XP_016866273.1 | |||
XM_017010785.1 | 153 | UTR 5 | XP_016866274.1 | |||
XM_017010786.1 | 153 | UTR 5 | XP_016866275.1 | |||
XM_017010787.1 | 153 | UTR 5 | XP_016866276.1 | |||
XM_017010788.1 | 153 | Intron | XP_016866277.1 |